Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Click to view available options
pH Range:
1 to 12
1.8 to 3.8
3.8 to 5.5
5.2 to 6.8
6 to 8.1
8 to 9.7
9.5 to 12
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use

Specifications
Specifications
pH Range | 1 to 12 |
Format | Strip |
For Use With (Application) | Routine pH testing |
Includes | Plastic supports, paper segments on each strip, complementary indicator charts |
Type | pH Indicator Test Strip, Integral Comparison Strip |
Dimensions (L x W) | 100 x 11 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction