Learn More
Description
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
Specifications
Specifications
| pH Range | 0 to 14 |
| Format | Strip |
| Includes | Color comparison chart |
| Quantity | 200 Pack |
| Type | Indicator Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.