Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ pH Indicators

Catalog No. 09924254 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
0 to 14
1 to 11
2 to 9
Quantity:
1 Roll
100 Pack
200 Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Quantity
09-924-254 2 to 9 100 Pack
09-924-255 1 to 11 1 Roll
09-924-253 0 to 14 100 Pack
09-924-250 0 to 14 200 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 09-924-254 Supplier Cytiva Supplier No. 10362010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.

Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.

  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
TRUSTED_SUSTAINABILITY

Specifications

pH Range 2 to 9
Format Strip
Includes Color comparison chart
Quantity 100 Pack
Type Indicator Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.