Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF

Catalog No. 0974450 Shop All Cytiva Products
Change view
Click to view available options
Filtration Area:
1100 cm2
260 cm2
535 cm2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtration Area
09-744-50 1100 cm2
09-744-45 260 cm2
09-744-53 535 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-744-50 Supplier Cytiva Supplier No. 2820
Add to Cart
Add to Cart

Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.

  • Three-layer filter permits filtration of larger volumes
  • Integrity testable
  • With inlet vent
  • Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0 μm polysulfone

Serum Prefilter Capsules. Retention (with air purge), < 2 mL

For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Description Polycap SPF 150 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 Pc.)
Filter Type Prefilter
Material (Membrane) Glass Microfiber, Polyethersulfone
Pore Size 1 μm
Filtration Area 1100 cm2
For Use With (Application) For Serum Prefiltration. Handle Tissue Culture Media, Nutrients, Immunologicals, Enzymes, Diagnostic Standards/Controls, Biologicals, Buffers, Virus Suspensions.
Housing Material Polypropylene
Inlet Connection (English) 1/2 in.
Inlet Style Stepped Barb
Max. Pressure 4.1 bar (60 psi)
Outlet Fitting 1/2 in.
Type Serum Prefilter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.