Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF
Click to view available options
Filtration Area:
1100 cm2
260 cm2
535 cm2
Description
- Three-layer filter permits filtration of larger volumes
- Integrity testable
- With inlet vent
- Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0μm polysulfone
Serum Prefilter Capsules. Retention (with air purge), <2mL
For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.

Specifications
Specifications
Autoclavable | Autoclavable |
Description | Polycap SPF 36 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 pc) |
Material (Membrane) | Glass Microfiber, Polyethersulfone |
Pore Size | 1 μm |
Filtration Area | 260 cm2 |
For Use With (Application) | For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions. |
Housing Material | Polypropylene |
Max. Pressure | 4.1 bar (60 psi) |
Outlet Style | Stepped Barb |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction