Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polycap Disposable Capsules, 75 AS

Catalog No. 09927842 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Outlet Style:
Female National Pipe Thread
Stepped Barb
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style Outlet Style Sterility
09-927-842 0.2 μm Female National Pipe Thread Female National Pipe Thread Sterile
09-927-231 0.45 μm Stepped Barb Stepped Barb Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-927-842 Supplier Cytiva Supplier No. 2706T
Add to Cart
Add to Cart

Polycap 75 AS filters for filtration of aqueous solutions

  • Sterile and nonpyrogenic
  • Integrity testable
  • With inlet vent
  • Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter

  • Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask

Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.

TRUSTED_SUSTAINABILITY

Specifications

Description Polycap AS 75 Capsule Filter, 0.2 μm, FNPT inlet and outlet (5 pcs)
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style Female National Pipe Thread
Max. Pressure 4.1 bar (60 psi)
Outlet Style Female National Pipe Thread
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.