Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polycap TF

Catalog No. 09924559 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-924-559 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-924-559 Supplier Cytiva Supplier No. 67103602
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytiva's Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.

  • Autoclavable Polycap TF capsule filters feature PTFE membranes with high chemical resistance
  • Common applications include venting, in-line filtration to sterilize and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters
  • Polycap TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
TRUSTED_SUSTAINABILITY

Specifications

Description Supported bidirectionally. Certain applications may require orientation, i.e., vents. Reverse flow only for low pressure applications.
Pore Size 0.2 μm
Housing Material Polypropylene
Outlet Style Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.