Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polydisc In-line Filter

Catalog No. 09744105 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
1.0 μm
5 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-744-105 1.0 μm
09-744-84 0.2 μm
09-927-227 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-744-105 Supplier Cytiva Supplier No. 67245000
Add to Cart
Edge
Add to Cart

For serum samples

  • Glass microfiber and polyethersulfone membranes
  • Contains stack of filter media
  • Radiation sterilized

TRUSTED_SUSTAINABILITY

Specifications

Material PES
Pore Size 1.0 μm
Flow Rate 500.0mL/min./cm sq.
For Use With (Application) For serum samples; In virology, microbiology, immunoassay
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.