Learn More
Description
Versatile, cost-effective filters designed for larger volume sample filtration
- Filter up to 1L with one device
- Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
- High-purity polypropylene housing maintains sample purity
- 50mm filter devices
- Autoclavable
- Lightweight (11.5g)
- For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
- Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
- Sterility cap on outlet (optional)
- No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
- Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
- Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe
Specifications
Specifications
| Autoclavable | Autoclavable |
| Description | Polydisc TF Filter, polypropylene, 0.45 μm (10 pcs) |
| Material (Membrane) | PTFE |
| Pore Size | 0.45 μm |
| Filtration Area | 16 cm2 |
| For Use With (Application) | Larger volume sample filtration |
| Housing Material | Polypropylene |
| Inlet Connection (Metric) | Tubing Nozzle 6 to 10 mm i.d. hose |
| Quantity | Pk. of 10 |
| Type | Polydisc TF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.