Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polydisc Inline Filters

Catalog No. 0974472 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
1 μm
Material (Membrane):
PES
PTFE
Polypropylene
Type:
Inline Filter
Polydisc AS
Polydisc TF
Quantity:
Pk. of 10
Pk. of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane) Type Quantity
09-744-72 1 μm Polypropylene Polydisc TF Pk. of 10
09-744-69 0.45 μm PTFE Polydisc TF Pk. of 10
09-744-66 0.2 μm PTFE Polydisc TF Pk. of 10
09-733-06 0.45 μm PES Polydisc AS Pk. of 50
09-744-90 0.45 μm PES Inline Filter Pk. of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 09-744-72 Supplier Cytiva Supplier No. 67215010
Add to Cart
Add to Cart

Versatile, cost-effective filters designed for larger volume sample filtration

  • Filter up to 1L with one device
  • Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
  • High-purity polypropylene housing maintains sample purity
  • 50mm filter devices
  • Autoclavable
  • Lightweight (11.5g)
Polydisc AS (Aqueous Solution)
  • For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
  • Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
  • Sterility cap on outlet (optional)
  • No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
Polydisc TF
  • Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
  • Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Description Polydisc TF Filter, polypropylene, 1.0 μm (10 pcs)
Material (Membrane) Polypropylene
Pore Size 1 μm
Filtration Area 16 cm2
For Use With (Application) Larger volume sample filtration
Housing Material Polypropylene
Inlet Connection (Metric) Tubing Nozzle 6 to 10 mm i.d. hose
Quantity Pk. of 10
Type Polydisc TF
Diameter (Metric) 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.