Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Polydisc TF In-line Filters
Click to view available options
Pore Size:
0.1 μm
Description
Available options for filtration media suit a range of aqueous and organic samples
- Hydrophobic PTFE membrane for filtration with chemically aggressive solutions, reagents and organic solvents
- High-purity polypropylene housing maintains sample purity
- 50mm filter
- Can be used in conjunction with a syringe or connected in-line (6-10mm I.D. hose) via barbed hose connections
- Autoclavable
- Lightweight (11.5g)
- Biosafe
- 1μm device features polypropylene prefilter for use with heavy contamination
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
For in-line filtration and isolation applications in pharmaceuticals, biotech research, laboratories and electronics
Compliance
Integrity testable with bubble point method

Specifications
Specifications
Description | Polydisc TF Filter, polypropylene, 0.1 μm (10 pcs) |
Material (Membrane) | PTFE |
Pore Size | 0.1 μm |
Filtration Area | 16 cm2 |
For Use With (Application) | For sample preparation of ground water samples for the analysis of dissolved heavy metals |
Housing Material | Polypropylene |
Inlet Connection (Metric) | Tubing Nozzle 6 to 10 mm i.d. hose |
Diameter (Metric) | 50 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction