Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ Whatman™ Qualitative Filter Paper Grade CF 12

Catalog No. 09920124
Click to view available options
Quantity:
100 Pk.

Whatman™ Grade CF 12 qualitative filter paper is ideal for general laboratory applications, such as removing particulate matter, clarifying liquids and separating solids from liquids in qualitative analysis. The Grade CF 12 offers medium retention and flow rate.

  • Qualitative filter paper sheets
  • Grade CF 12
  • Ideal for general filtration applications
  • Medium retention and flow rate
  • Suitable for various industries including pharmaceutical, environmental and food testing
TRUSTED_SUSTAINABILITY

Specifications

Description Qualitative filter paper
Product Line Whatman™
Product Type Qualitative Filter Paper
Quantity 100 Pk.
Material Cellulose fibers
Format Sheets 210 x 297 mm
Purity or Quality Grade Grade CF 12
For Use With (Application) Suitable for various industries including pharmaceutical, environmental and food testing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.