Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Reeve Angel 230 Creped Qualitative Circles

Catalog No. 0980517B Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
9 cm
11 cm
12.5 cm
15 cm
18.5 cm
20 cm
24 cm
25 cm
33 cm
40 cm
50 cm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
09-805-17B 18.5 cm
52-302-00 20 cm
09-805-17D 33 cm
WTI-230-25 25 cm
WTI-230-11 11 cm
WTI-230-12-5 12.5 cm
WTI-230-24 24 cm
09-803A 9 cm
09-803D 15 cm
05-713-507 40 cm
05-713-508 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 09-805-17B Supplier Cytiva Supplier No. 5230185
Add to Cart
Edge
Add to Cart

Provide reliable performance for non-critical industrial laboratory and student lab applications

Retain coarse particles.

  • Made from cellulose
  • Creped surface for fast filtration of coarse particles
  • Particle retention: 25 to 30μm
  • Thickness: 0.40mm

Should not be used for kjeldahl nitrogen analysis

TRUSTED_SUSTAINABILITY

Specifications

Description 230 18.5 cm 50/pk
Format Disc
Material Cellulose
Particle Retention 25 to 30 μm
For Use With (Application) A general qualitative filter with creped surface. High filtration speed and high loading capacity. Widely used in industrial laboratories and students laboratories for routine filtration.
Grade 230
Porosity Coarse
Quantity 50/Pk.
Thickness (Metric) 0.4 mm
Dimensions (L x W) 18.50 cm
Diameter (English) 7.28 in.
Diameter (Metric) 18.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.