Learn More
Cytiva Whatman™ Syringe-Type Membrane Holder
Catalog No. 09924549 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inside:
13 mm
25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside
09-924-549 25 mm
09-908-1 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-924-549 Supplier Cytiva Supplier No. 1980002
Add to Cart
Add to Cart
Stainless steel membrane holder for use with a standard syringe
  • Designed for the quick and easy clarification, sterilization, and removal of particulates from small volume samples (typically for HPLC applications)
  • holders contain PTFE gaskets and O-rings and allow the membrane to be autoclaved in place without the filter sticking to the holder.
  • Luer lock fittings connect to a standard syringe and offer convenience and ease of use for clarification, sterilization, and removal of particulates from small volumes of liquid
  • TRUSTED_SUSTAINABILITY
    Specifications
    Material Stainless Steel
    Diameter (Metric) Inside 25 mm