Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL

Catalog No. 09927872 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
1.2 μm
10 to 12 μm
25 to 30 μm
Color:
Clear
Natural
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Color
09-927-872 1.2 μm Natural
05-713-849 25 to 30 μm Clear
09-927-350 10 to 12 μm Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-927-872 Supplier Cytiva Supplier No. 77009901
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis

  • Natural polypropylene plates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Description The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
Array 4 x 6
Pore Size 1.2 μm
Color Natural
Material Polypropylene
No. of Wells 24
Sterility Non-sterile
Well Shape Filter
For Use With (Application) Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
Includes Long drip director and Glass Microfiber GF/C Filter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.