Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UNIPLATE™ Microplates

Catalog No. PF77015102 Shop All Cytiva Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
5 mL
10 mL
Well Shape:
Flat Bottom (F-well)
Round
No. of Wells:
24
48
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Well Well Shape No. of Wells
PF77015102 Non-sterile 10 mL Round 24
PF77015500 Non-sterile 5 mL Flat Bottom (F-well) 48
PF77015110 Sterile 10 mL Round 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PF77015102 Supplier Cytiva Supplier No. 77015102
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

  • Withstand low temperatures (-80°C) for long-term storage
  • Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Description UNIPLATE microplates
Array 4 x 6
Color Natural
Material Polypropylene
Volume (Metric) Well 10 mL
No. of Wells 24
Sterility Non-sterile
Well Shape Round
For Use With (Application) Ideal for homogeneous assay techniques utilized in high-throughput screening
Quantity 25/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.