Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ 60mL BOD Bottles with Glass Pennyhead Stoppers
SDP

Catalog No. 02926100B Shop All DWK Life Sciences Products
Click to view available options
No. Sequence:
1 to 36
37 to 72
73 to 108
Specials
Unnumbered

Securely store samples with these BOD bottles, which include pennyhead-style stoppers

  • Type I borosilicate glass construction
  • Height: 115mm (4.53 in.)
  • Diameter: 43mm (1.69 in.)
  • Capacity: 60mL (2 oz.)

TRUSTED_SUSTAINABILITY

Specifications

Barcode Yes
Volume (Metric) 60 mL
For Use With (Application) Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Marking Spot Yes
Material Borosilicate Glass
No. Sequence 1 to 36
Stopper Pennyhead
Volume (English) 2.02 oz.
Height (English) 4.5 in.
Height (Metric) 11.5 cm
Diameter (English) 1.69 in.
Diameter (Metric) 4.3 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.