Learn More
DWK Life Sciences Wheaton™ ABC Vial Caps
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
Specifications
Specifications
Material (Liner) | PTFE/Silicone |
Color | Red |
Material | Polypropylene |
Closure Size | 9 mm |