Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
SDP

Catalog No. 02911378 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk pack
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Packaging
02-911-378 1000 mL Valu-bulk pack
02-911-350 30 mL Convenience pack
02-911-351 60 mL Convenience pack
02-911-352 125 mL Convenience pack
02-911-353 250 mL Convenience pack
02-911-354 500 mL Convenience pack
02-911-355 1000 mL Convenience pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 02-911-378 Supplier DWK Life Sciences Supplier No. W216864
Only null left
Add to Cart
Add to Cart

Amber Type III soda lime glass protects light-sensitive solutions

  • Available in partitioned Convenience Pack or Valu-Bulk™ cartons
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

Order Info

Caps can be ordered separately.

Compliance

USP Type III soda-lime glass

TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Amber
Diameter (English) 3.7 in.
Material Soda Lime Glass
Packaging Valu-bulk pack
Closure Color White
Height (Metric) 208 mm
Height (English) 8.18 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 94 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.