Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Capillary Tubes
SDP

Catalog No. p-3208247
Click to view available options
Volume (Metric):
1 to 5 μL
10 to 50 μL
100 to 200 μL
5 to 25 μL
Color:
Green
Red
White

Glass Capillaries for Acura 841 Positive Displacement Micropipette

TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
Color White
Volume (Metric) 1 to 5 μL
Type Capillary Tube
For Use With (Equipment) Socorex Positive Displacement Micropipette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.