Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
SDP

Catalog No. 02911325 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk pack

Ideal for solvent, chemical and sample storage

  • Narrow mouth enables liquids to be carefully poured to prevent spillage
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

  • Available in partitioned Convenience or Valu-Bulk™ cartons

Order Info

Caps can be ordered separately.

Compliance

USP Type-III soda-lime glass

TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Clear
Diameter (English) 3.7 in.
Material Soda Lime Glass (Type 3)
Packaging Convenience pack
Closure Color White
Certifications/Compliance EPA, FDA
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 208 mm
Product Type Boston Round Bottle
Height (English) 8.18 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 94 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.