Learn More
DWK Life Sciences Wheaton™ Connection Connectors
SDP
Safely joins two exterior threaded glass components without hooks, springs or clamps
$346.50
Specifications
Certifications/Compliance USP Type I, ASTM E438 Type I, Class A
For Use With (Application) Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
For Use With (Equipment) Micro soxhlet extraction apparatus
Includes Screw cap
Material Borosilicate Glass, Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Thread Size Price Quantity & Availability  
Catalog Number Mfr. No. Thread Size Price Quantity & Availability  
15-320-19E
SDP
View Documents
DWK Life Sciences
125488
20-400 and 24-400
Case of 6 for $346.50
Only null left
Add to Cart
Description
Description
  • Screw cap configuration creates a grease-free, vacuum- tight seal
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Available with white polypropylene or black phenolic caps
  • Phenolic connectors have a working temperature of up to 180
Specifications
Specifications
USP Type I, ASTM E438 Type I, Class A
Micro soxhlet extraction apparatus
Borosilicate Glass, Polypropylene
Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Screw cap
Screw Thread Connector
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.