Description
- Conforms to USP and ASTM requirements
- 38, 45 or 51mm screw head
- 2.6mm wall thickness
- Bottle material made of borosilicate glass
- Autoclavable
- Reusable
Specifications
Specifications
Height (Metric) | 355 mm |
Diameter (Metric) | 110 mm |