Learn More
DWK Life Sciences Disposable 10μL Pipette Microtips
SDP
Catalog No. p-161869
Click to view available options
Sterility:
Non-sterile
Tip Style:
Short
Standard Length
Quantity:
960
1000
Premium pipette tips covering most research and routine applications, compatible with most pipettes
  • For both fixed- and variable-volume models
  • Precision injection molding care for air-tight fit
  • Made from high-grade, virgin polypropylene
  • Free from trace metals for excellent accuracy and reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
For Use With (Application) Premium pipette tips covering most research and routine applications, compatible with most pipettes, excellent accuracy and reproducibility
Format Rack
Tip Style Standard Length
Quantity 960
Disposable Single-use
Material Virgin Polypropylene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.