Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Hybridization Bottles
SDP

Catalog No. 1324597B Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric):
75 mm
100 mm
150 mm
300 mm
Diameter (Metric) Inner:
35 mm
70 mm
Product Type:
Hybridization Bottle
Safety-coated Hybridization Bottle
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Product Type
13-245-97B 100 mm 35 mm Hybridization Bottle
13-245-97C 150 mm 35 mm Hybridization Bottle
13-245-97A 75 mm 35 mm Hybridization Bottle
13-245-97D 300 mm 35 mm Hybridization Bottle
13-245-97E 300 mm 70 mm Hybridization Bottle
13-245-97J 300 mm 35 mm Safety-coated Hybridization Bottle
13-245-97F 75 mm 35 mm Safety-coated Hybridization Bottle
13-245-97G 100 mm 35 mm Safety-coated Hybridization Bottle
13-245-97H 150 mm 35 mm Safety-coated Hybridization Bottle
13-245-97K 300 mm 70 mm Safety-coated Hybridization Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 13-245-97B Supplier DWK Life Sciences Supplier No. 805001
Add to Cart
Add to Cart

For use in nucleic acid and protein blotting procedures that require a minimum of probe solution

  • Manufactured from heavy-wall borosilicate glass
  • Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
  • Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
  • Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage

Order Info

Items come as one per case

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) Inner 1.37 in.
Material Borosilicate Glass
Diameter (Metric) Inner 35 mm
Closure Type Screw Cap
Product Type Hybridization Bottle
Closure Size 45 mm
Length (Metric) 100 mm
Coating Plastisol
For Use With (Application) Nucleic acid and protein blotting procedures that require a minimum of probe solution
Liner Material PTFE or Silicone
Certifications/Compliance USP Type I
Length (English) 3.93 in.
Closure Material Polycarbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.