Learn More
Description
- Manufactured from heavy-wall borosilicate glass
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
- Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
Order Info
Items come as one per case
Specifications
Specifications
| Closure Type | Screw Cap |
| Product Type | Safety-coated Hybridization Bottle |
| Diameter (English) Inner | 1.37 in. |
| Material | Borosilicate Glass |
| Diameter (Metric) Inner | 35 mm |
| Closure Size | 45 mm |
| Length (Metric) | 100 mm |
| Certifications/Compliance | USP Type I |
| Coating | Plastisol |
| Length (English) | 3.93 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
