Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Lab 45 Graduated Safety-Coated Bottles
GSA_VASDP

Catalog No. 06451293 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL

Use as media or reagent bottle

Plastisol coated for safety

  • Made from clear borosilicate glass
  • Attached 45mm white polypropylene screw cap
  • Pour ring to eliminate drips
  • Autoclavable
  • Color-coded caps available separately

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 17 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Clear
Diameter (English) 3.38 in.
Includes Wedge shaped inner sealing ring and Polypropylene pouring ring
Material Borosilicate Glass
Closure Color White
Graduated Yes
Height (Metric) 186 mm
Height (English) 7.32 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 86 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.