Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (13mm)
SDP

Catalog No. p-3306211 Shop All DWK Life Sciences Products
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.

13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material LDPE
No. per Case 1000 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.