Learn More
DWK Life Sciences Wheaton™ Magnetic Stirrers Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  DWK Life Sciences W900701B
 View more versions of this product
Catalog No. 02-912-638
  Check Availability
Add to cart
Description
Description
  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100VAC Japan
5 to 200RPM
Magnetic Stirrers