Learn More
DWK Life Sciences Wheaton™ Micro Kit and Accessories
SDP
Catalog No. 1130232C Shop All DWK Life Sciences Products
Click to view available options
Product Type:
Claisen Adapter
Connecting Adapter
WHEATON standard taper Clear-Seal™ grease-free joints provide smooth, unground surfaces, which seal without grease and reduce seizing and contamination from grease
  • Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately
  • Clear-Seal™ joints can replace ground units in any application and provide other important advantages over ground joints
  • Superior in vacuum and radioactive applications
  • Smooth surfaces resist contamination
  • Cleaning is simple and thorough
  • Interchangeable with ground joints
  • All chemical changes and thermometer scales remain visible through the joints
  • Clear-Seal joints have never been weakened by grinding and are formed from special wall tubing that is heavier than standard wall for extra strength
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
  • Clear-Seal joints are produced to American Standard Taper dimensions and can be used interchangeably with any standard taper ground joints
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Wheaton micro kit
Includes 19/22 Clear-Seal™ joints
Product Type Claisen Adapter
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.