Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Natural Polypropylene Caps for Narrow-Mouth Containers
SDP

Catalog No. 0987712A Shop All DWK Life Sciences Products
Click to view available options
Neck Finish:
13-425
20-410
24-410
38-430

For use with Wheaton leak-resistant narrow mouth bottles

  • Leak-proof design
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 24-410
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.