Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
SDP

Catalog No. p-76237 Shop All DWK Life Sciences Products
Click to view available options
Color:
Black
White
Product Type:
Cap
Open Top Cap

For use with Wheaton Glass Diagnostic Bottles

  • Unlined
  • Open- and closed-top caps
  • 20-400 screw thread
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polypropylene
No. per Case 300/Cs.
Neck Finish 20-400
Certifications/Compliance ISO 9001
Product Type Open Top Cap
For Use With (Equipment) Screw neck diagnostic bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.