Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Orange
SDP

Catalog No. p-2429796 Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100 Cs.
1000 Cs.

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 20-410
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.