Learn More
DWK Life Sciences Wheaton™ Polypropylene Clamps
SDP
For Erlenmeyer Shake Flasks
Supplier:  DWK Life Sciences WPSFC0250
 View more versions of this product
Catalog No. 02-924-986
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Carefully engineered to provide both clipping elasticity and motion resistance
  • Feature a non-scratch design
  • Offered in a standard blue color accompanied by stainless-steel screws and washers
  • Fit all industry standard shaking incubators
  • Autoclavable for reuse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL Shaker Flask
Polypropylene
1 Ea.
Plastic Shaker Flask Clamp
Blue
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.