Learn More
DWK Life Sciences Wheaton™ Polypropylene Clamps
For Erlenmeyer Shake Flasks
Supplier: DWK Life Sciences WPSFC0500
Description
- Carefully engineered to provide both clipping elasticity and motion resistance
- Feature a non-scratch design
- Offered in a standard blue color accompanied by stainless-steel screws and washers
- Fit all industry standard shaking incubators
- Autoclavable for reuse.
Specifications
| 500 mL Shaker Flask | |
| Polypropylene | |
| 1 Ea. |
| Plastic Shaker Flask Clamp | |
| Blue |