Learn More
DWK Life Sciences Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers
Shop All DWK Life Sciences Products

Description
- Leak-proof design
- Autoclavable
Specifications
Specifications
| Color | Red |
| Material | Polypropylene |
| No. per Case | 72 Cs. |
| Neck Finish | 13-425 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.