Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
SDP

Catalog No. 03410733 Shop All DWK Life Sciences Products
Click to view available options
Septum:
PTFE/Butyl
PTFE/Silicone
Color (Seal):
Natural
Red/White
White/Transparent Blue

Screw thread finish eliminates the need for crimping tools

  • For use with CTC Analytics Systems requiring magnetic closures
  • Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
TRUSTED_SUSTAINABILITY

Specifications

Color (Seal) Natural
Material Butyl, PTFE
Septum PTFE/Butyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.