Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Serum Bottles and Vials
GSA_VASDP

Catalog No. p-45371 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
60 mL
100 mL
125 mL
Color:
Amber
Clear

These bottles have exceptional wall strength and superior heat and chemical tolerances

Ideal for long- and short-term sample storage and vaccine/injectable drug containers.

  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage

Compliance

Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.1 oz., 2 mL
Capacity (Metric) 2 mL
Capacity (English) 0.1 oz.
Color Clear
Diameter (English) 0.61 in.
Material Borosilicate Glass
Height (Metric) 36 mm
Height (English) 1.41 in.
Diameter (Metric) 15.6 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.