Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Serum Vials
SDP

Catalog No. 06446G Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
2 mL
3 mL
5 mL
10 mL
10.0 mL
20 mL
Color:
Amber
Clear
Diameter (Metric) Outer:
15 mm
17 mm
22 mm
24 mm
30.5 mm

Available as clear and amber vials

Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.

  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination

Lyophilization, packaging, and sampling

Compliance

Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7 mm
Color Amber
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 2 mL
For Use With (Application) For lyophilization, packaging, sampling and freeze drying applications
Volume (English) 0.5 dr.
Autoclavable Autoclavable
Closure Type Crimp Cap
Height (Metric) 32 mm
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.