Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
SDP

Catalog No. 0298445 Shop All DWK Life Sciences Products
Click to view available options
Neck Finish:
22-400
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
89-400
No. per Case:
144 Cs.
48 Cs.
72 Cs.

Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products

  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 53-400
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Round bottle
Closure Liner Polyvinyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.