Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqua Phoenix™ Where's the Water Deluxe Kit Components
Shop All Fisherbrand Products

Click to view available options
Product Type:
Beaker
Medicine cups
Mini plastic animals
Mixing bowl
Tacky putty
Description
Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications
Specifications
Product Type | Beaker |
Grade | K to 5 |
For Use With (Application) | Education Kit |
Material | HDPE |
Capacity (Metric) | 1000 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction