Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ White and Black 384-Well Immuno Plates
GREENER_CHOICE

Catalog No. p-4555270
Click to view available options
Color:
Black
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.

Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.

Features:

  • Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Plate is offered as black or white for optimal choice of application type
  • Certified binding offers homogeneity/reproducibility.

White:

  • Maximum reflection/minimum autofluorescence and autoluminescence

Black:

  • Minimizes crosstalk, back-scattered light and background in fluorescence reading

Compatible with:

Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering information:

Availability may vary by country

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Hydrophilic
Format Standard
Lid Without Lid
Detection Method Chemiluminescence
Array 16 x 24
Packaging 30 Cases
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Fluorescence and Luminescence
No. per Case 30
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F384
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.