Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Lab Coat, Series 4440

Catalog No. 17624128 Shop All 3M Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Knitted cuffs.
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Single-use
Model 4440-L
Seam Type Serged
Size (English) Chest 36 to 38 in.
Size (Metric) Chest 91 to 97 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Catering, Cleanrooms, Electronics Manufacturing, General Maintenance, Laboratories
Weight (English) 1.93 oz.
Construction Microporous Laminate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.