Learn More
Eisco™ White Quartzite, Metamorphic Rock Specimen - Hand Sample - Approx. 3 Inch
SCI_ED
Catalog No. S02918 Shop All Eisco Products
Change view
Click to view available options
Specimen Size:
3 in.
Catalog No. Specimen Size
S02918 3 in.
1 options
Catalog No. S02918 Supplier Eisco™ Supplier No. ESNG0074HAND
Add to Cart
Add to Cart
Quartzite is formed from sandstone that has been tightly compressed to the point where the clasts have recrystallized to form a new rock. This formation mechanism, coupled the inherent strength of quartz, makes quartzite a very durable rock.
  • Quartzite is formed from sandstone that has been tightly compressed to the point where the clasts have recrystallized to form a new rock
  • This formation mechanism, coupled the inherent strength of quartz, makes quartzite a very durable rock
  • Useful in the classroom for lessons on metamorphic rocks
  • Ideal for identification and classification exercises
  • While each specimen will vary slightly in shape, size and color, it will measure approximately 1 inch in length
  • The specimens are packaged in a transparent bag that is labeled and barcoded for easy identification and storage
  • Each specimen is selected and hand processed by an actual geologist, ensuring that each sample exhibits defining features
  • Rocks and minerals are sourced from a variety of locations around the world and are processed by hand in Victor, NY.
Specifications
Product Type Metamorphic Rock
Specimen Size 3 in.