Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ White SMS Coveralls with Elastic Wrists and Open Ankles

Catalog No. 17184220
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large

SMS coveralls provide three layers of protection against particulates such as dirt and dust, and light liquid splashes or sprays. Lightweight and breathable fabric allows heat to escape so wearers stay cool.

  • SMS coveralls are lightweight and breathable, so wearers stay cool throughout long shifts
  • Durable, three-layer SMS fabric (Spunbond Meltblown Spunbond) protects against light liquid splashes and sprays and provides an ideal particulate barrier
  • Storm flap covers the zipper to help keep non-hazardous particles and liquids from entering zipper seams
  • Wider, gusseted leg area gives wearer more room to move in all directions without ripping fabric
  • Elastic wrists offer a secure fit to help keep sleeves in place to prevent skin exposure
  • Elastic back stretches with user for more comfort and better tear resistance
  • Possible applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle
Latex Free Yes
Model 2222
No. of Pockets None
For Use With (Application) Asbestos and Lead Abatement, Mold Remediation, Manufacturing, Assembly Operations
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.