Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whitney™ Medical Solutions Safe-Keeper™ Biohazard Container

Catalog No. p-36349 Shop All Fisherbrand Products
Click to view available options
Capacity (English):
1100 cc
2250 cc
Height (English):
0.75 in.
11 in.
Length (English):
2 in.
5 in.

Catalog No. 14-375-269

Only null left
Add to Cart
Add to Cart

Space-saving, economical, disposable point of use contaminated waste containers for disposal of capillary tubes, pipette tips, etc.

  • For disposal of capillary tubes, pipette tips, stir rods and other point-of-use items
  • Maximize workspace with hanging biohazard disposal units
  • Made of sturdy plastic-lined paperboard
  • Solid-bottom design improves rigidity and waste capacity
  • Safe-Keeper can be autoclaved or incinerated
  • Accommodates up to 1100cc of material and comes with a Hook-Up™ self-adhesive hanger
  • Made in USA

Specifications

Capacity (English) 2250 cc
Dimensions (L x W x H) 2 x 2 x 0.75 in. (5.1 x 5.1 x 1.9 cm)
Height (Metric) 1.9 cm
Length (Metric) 5.1 cm
Width (Metric) 5.1 cm
Autoclavable Autoclavable
Liner Material Plastic
For Use With (Application) Disposal of capillary tubes, pipette tips, stir rods and other point-of-use items
Width (English) 2 in.
Length (English) 2 in.
Height (English) 0.75 in.
Material Paperboard
Product Type Self-adhesive Hook
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.