Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Whole Serum Polyclonal Antibody
For Immunostaining of cell suspensions.
Supplier: MP Biomedicals™ 0856009
Description
- Used for acetone-fixed frozen tissue sections, and formalin-fixed, paraffin-embedded tissue sections
- Forensic antiserum
- Anti-bear

Specifications
Immunoassay | |
Whole serum | |
2 mL | |
Store at 2 to 8°C |
Polyclonal | |
Goat | |
Primary | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction