Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WHSC1 Antibody (CL1063), Novus Biologicals™
SDP

Catalog No. NB395328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395328 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395328 Supplier Novus Biologicals Supplier No. NBP25296325UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WHSC1 Monoclonal antibody specifically detects WHSC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen WHSC1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL1063
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias EC 2.1.1.43, FLJ23286, IL5 promoter REII region-binding protein, KIAA1090, MGC176638, MMSETNSD2Multiple myeloma SET domain-containing protein, multiple myeloma SET domain containing protein type III, nuclear receptor binding SET domain protein 2, Nuclear SET domain-containing protein 2, probable histone-lysine N-methyltransferase NSD2, Protein trithorax-5, REIIBP, trithorax/ash1-related protein 5, TRX5, WHS, Wolf-Hirschhorn syndrome candidate 1, Wolf-Hirschhorn syndrome candidate 1 protein
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SANGKTPSCEVNRECSVFLSKAQLSSSLQEGVMQKFNGHDALPFIPADKLKDLTSRVFNGEPGAHDAKLRFESQEMKGIGTPPNTTPIKNGSPEIKLKITKTYMNGKPLFESSICGD
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7468
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.