Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WHSC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567544 Supplier Invitrogen™ Supplier No. PA567544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WHSC2 Polyclonal Antibody for Western Blot, IHC (P)

This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WHSC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NELFA
Gene Accession No. Q8BG30, Q9H3P2
Gene Alias FLJ10442; FLJ25112; LOW QUALITY PROTEIN: negative elongation factor A; mWHSC2; Negative elongation factor A; negative elongation factor complex member A; negative elongation factor complex member A, Whsc2; Nelfa; NELF-A; P/OKcl.15; Whsc2; Whsc2h; Wolf-Hirschhorn syndrome candidate 2; Wolf-Hirschhorn syndrome candidate 2 homolog; Wolf-Hirschhorn syndrome candidate 2 protein
Gene Symbols NELFA
Host Species Rabbit
Immunogen Fusion protein of human WHSC2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24116, 7469
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.