Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WHSC2 Polyclonal Antibody

WHSC2 Polyclonal Antibody
Supplier: Thermo Scientific PA5115637
Description
WHSC2 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| WHSC2 | |
| Polyclonal | |
| Unconjugated | |
| NELFA | |
| FLJ10442; FLJ25112; LOW QUALITY PROTEIN: negative elongation factor A; mWHSC2; Negative elongation factor A; negative elongation factor complex member A; negative elongation factor complex member A, Whsc2; Nelfa; NELF-A; P/OKcl.15; Whsc2; Whsc2h; Wolf-Hirschhorn syndrome candidate 2; Wolf-Hirschhorn syndrome candidate 2 homolog; Wolf-Hirschhorn syndrome candidate 2 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24116, 7469 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8BG30, Q9H3P2 | |
| NELFA | |
| A synthesized peptide derived from human NELFA(Accession Q9H3P2), corresponding to amino acid residues Q187-N237. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction